2-Methyltetrahydrofuran, 99+%, Extra Dry, stabilized, AcroSeal™, Thermo Scientific Chemicals
2-Methyltetrahydrofuran, 99+%, Extra Dry, stabilized, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Methyltetrahydrofuran, 99+%, Extra Dry, stabilized, AcroSeal™, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
100 mL
2.5 L
Packaging:
AcroSeal™ Glass Bottle
Catalog number 396620010
Price (USD)/ Each
185.65
Online exclusive
206.00 
Save 20.35 (10%)
-
Add to cart
Quantity:
1 L
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (USD)/ Each
185.65
Online exclusive
206.00 
Save 20.35 (10%)
Add to cart
2-Methyltetrahydrofuran, 99+%, Extra Dry, stabilized, AcroSeal™, Thermo Scientific Chemicals
Catalog number396620010
Price (USD)/ Each
185.65
Online exclusive
206.00 
Save 20.35 (10%)
-
Add to cart
Chemical Identifiers
CAS96-47-9
IUPAC Name2-methyloxolane
Molecular FormulaC5H10O
InChI KeyJWUJQDFVADABEY-UHFFFAOYNA-N
SMILESCC1CCCO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Peroxides=<50 ppm
Infrared spectrumConforms
GC>=99.0 %
Appearance (Form)Liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only