Thermo Scientific Chemicals

1-Hexanesulfonic acid, sodium salt, 99+%, Ion pair chromatography, anhydrous, Thermo Scientific Chemicals

Catalog number: 396671000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

1-Hexanesulfonic acid, sodium salt, 99+%, Ion pair chromatography, anhydrous, Thermo Scientific Chemicals

Catalog number: 396671000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 396671000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2832-45-3
IUPAC Name
sodium hexane-1-sulfonate
Molecular Formula
C6H13NaO3S
InChI Key
QWSZRRAAFHGKCH-UHFFFAOYSA-M
SMILES
[Na+].CCCCCCS([O-])(=O)=O
Appearance (Form)
Crystalline solid
UV
at 210 nm A: =<0.1
UV
at 230 nm A: =<0.04
Suitability
(HPLC gradient) Passes test
Appearance (Color)
White

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ion pair chromatography
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text