Thermo Scientific Chemicals

1-Heptanesulfonic acid, sodium salt, 99+%, Ion pair chromatography, anhydrous, Thermo Scientific Chemicals

Catalog number: 396680250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

1-Heptanesulfonic acid, sodium salt, 99+%, Ion pair chromatography, anhydrous, Thermo Scientific Chemicals

Catalog number: 396680250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 396680250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
22767-50-6
IUPAC Name
sodium heptane-1-sulfonate
Molecular Formula
C7H15NaO3S
InChI Key
REFMEZARFCPESH-UHFFFAOYSA-M
SMILES
[Na+].CCCCCCCS([O-])(=O)=O
Appearance (Form)
Crystalline powder
UV
(0.5 M in water)
UV
at 230 nm A: =<0.04
Appearance (Color)
White
Loss on drying
=<2 % (150°C)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ion pair chromatography
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text