1-Heptanesulfonic acid, sodium salt, 99+%, Ion pair chromatography, anhydrous, Thermo Scientific Chemicals
1-Heptanesulfonic acid, sodium salt, 99+%, Ion pair chromatography, anhydrous, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Heptanesulfonic acid, sodium salt, 99+%, Ion pair chromatography, anhydrous, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number 396681000
Price (USD)/ Each
865.65
Online exclusive
962.00 
Save 96.35 (10%)
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
865.65
Online exclusive
962.00 
Save 96.35 (10%)
Add to cart
1-Heptanesulfonic acid, sodium salt, 99+%, Ion pair chromatography, anhydrous, Thermo Scientific Chemicals
Catalog number396681000
Price (USD)/ Each
865.65
Online exclusive
962.00 
Save 96.35 (10%)
-
Add to cart
Chemical Identifiers
CAS22767-50-6
IUPAC Namesodium heptane-1-sulfonate
Molecular FormulaC7H15NaO3S
InChI KeyREFMEZARFCPESH-UHFFFAOYSA-M
SMILES[Na+].CCCCCCCS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
UV(0.5 M in water)
UVat 230 nm A: =<0.04
Appearance (Color)White
Loss on drying=<2 % (150°C)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ion pair chromatography
RUO – Research Use Only