Imidazole, ACS reagent, Thermo Scientific Chemicals
Imidazole, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Imidazole, ACS reagent, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 396745000
Price (USD)/ Each
154.65
Online exclusive
172.00 
Save 17.35 (10%)
-
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
154.65
Online exclusive
172.00 
Save 17.35 (10%)
Add to cart
Imidazole, ACS reagent, Thermo Scientific Chemicals
Catalog number396745000
Price (USD)/ Each
154.65
Online exclusive
172.00 
Save 17.35 (10%)
-
Add to cart
Chemical Identifiers
CAS288-32-4
IUPAC Name1H-imidazole
Molecular FormulaC3H4N2
InChI KeyRAXXELZNTBOGNW-UHFFFAOYSA-N
SMILESN1C=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
pH9.5 to 11.0 (5% at 25°C)
Appearance (Form)Powder or crystals
Titration with HClO4>=99 %
Residue after ignition=<0.1 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only