1x1 image pixel for data collection
Poly(vinyl alcohol), 88% hydrolyzed, average M.W. 20000-30000
Poly(vinyl alcohol), 88% hydrolyzed, average M.W. 20000-30000
Poly(vinyl alcohol), 88% hydrolyzed, average M.W. 20000-30000
Poly(vinyl alcohol), 88% hydrolyzed, average M.W. 20000-30000
Thermo Scientific Chemicals

Poly(vinyl alcohol), 88% hydrolyzed, average M.W. 20000-30000

CAS: 9002-89-5 | (C2H4O)n | 44.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
25 g
500 g
1 kg
Packaging:
View all
Plastic bottle
Catalog number 396765000
Price (USD)
94.65
Online Exclusive
105.00
Save 10.35 (10%)
Each
In stock
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
94.65
Online Exclusive
105.00
Save 10.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS9002-89-5
Molecular Formula(C2H4O)n
SMILESOC(-*)C-*
MDL NumberMFCD00081922
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Almost white to light yellow
Appearance (Form)Powder
Infrared spectrumConforms
Degree of hydrolysis87.0 to 89.0 %
Volatile matter=<5.0 % (105°C, 3 h)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0449225Certificate of AnalysisJul 11, 2025396760010, 396760250, 396765000, 396760025
A0460769Certificate of AnalysisMar 20, 2024396760010, 396760250, 396765000, 396760025
A0449225Certificate of AnalysisMar 20, 2024396760010, 396760250, 396765000, 396760025
A0396259Certificate of AnalysisDec 14, 2022396760010, 396760250, 396765000, 396760025
A0413696Certificate of AnalysisDec 13, 2022396760010, 396760250, 396765000, 396760025
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection