Thermo Scientific Chemicals

Methyl 4-(aminomethyl)benzoate hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: 396780050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Methyl 4-(aminomethyl)benzoate hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: 396780050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 396780050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6232-11-7
IUPAC Name
methyl 4-(aminomethyl)benzoate hydrochloride
Molecular Formula
C9H12ClNO2
InChI Key
GIZCKBSSWNIUMZ-UHFFFAOYSA-N
SMILES
Cl.COC(=O)C1=CC=C(CN)C=C1
Appearance (Color)
White
Appearance (Form)
Crystalline powder or crystals
Titration Argentometric
96.0 to 104.0 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text