Thermo Scientific Chemicals

1-Propanol, 99.5%, Thermo Scientific Chemicals

Catalog number: 396941000
100 mL, Each
Thermo Scientific Chemicals

1-Propanol, 99.5%, Thermo Scientific Chemicals

Catalog number: 396941000
100 mL, Each
Quantity
Catalog number: 396941000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
71-23-8
IUPAC Name
propan-1-ol
Molecular Formula
C3H8O
InChI Key
BDERNNFJNOPAEC-UHFFFAOYSA-N
SMILES
CCCO
Appearance (Color)
Clear colorless
Filtered to 0.2 micrometer
Filtered
Appearance (Form)
Liquid
Water
=<0.005 % (Coulometric)
GC
>=99.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text