Thermo Scientific Chemicals

Chlorobenzene, 99.8%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 396970010
1 L, AcroSeal™ Glass Bottle, Each
Thermo Scientific Chemicals

Chlorobenzene, 99.8%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 396970010
1 L, AcroSeal™ Glass Bottle, Each
Quantity
Packaging
Catalog number: 396970010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
108-90-7
IUPAC Name
chlorobenzene
Molecular Formula
C6H5Cl
InChI Key
MVPPADPHJFYWMZ-UHFFFAOYSA-N
SMILES
ClC1=CC=CC=C1
Appearance (Form)
Liquid
GC
>=99.8 %
Appearance (Color)
Clear colorless
Water
=<0.005 % (Coulometric)
Filtered to 0.2 micrometer
Filtered

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text