Thermo Scientific Chemicals

3,4-Diaminoanisole dihydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: 396990010
1 g, Each
Thermo Scientific Chemicals

3,4-Diaminoanisole dihydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: 396990010
1 g, Each
Quantity
Catalog number: 396990010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
59548-39-9
IUPAC Name
4-methoxybenzene-1,2-diamine hydrochloridyl
Molecular Formula
C7H10ClN2O
InChI Key
NOFDEJFDTTYTCG-UHFFFAOYSA-N
SMILES
[Cl].COC1=CC=C(N)C(N)=C1
Appearance (Color)
Violet
Titration Argentometric
>=97.5 to =<102.5 %
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text