3,4-Diaminoanisole dihydrochloride, 98%, Thermo Scientific Chemicals
3,4-Diaminoanisole dihydrochloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,4-Diaminoanisole dihydrochloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Catalog number 396990250
Price (USD)/ Each
370.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
370.00
Add to cart
3,4-Diaminoanisole dihydrochloride, 98%, Thermo Scientific Chemicals
Catalog number396990250
Price (USD)/ Each
370.00
-
Add to cart
Chemical Identifiers
CAS59548-39-9
IUPAC Name4-methoxybenzene-1,2-diamine hydrochloridyl
Molecular FormulaC7H10ClN2O
InChI KeyNOFDEJFDTTYTCG-UHFFFAOYSA-N
SMILES[Cl].COC1=CC=C(N)C(N)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Violet
Titration Argentometric>=97.5 to =<102.5 %
Appearance (Form)Crystalline powder
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only