Thermo Scientific Chemicals

4-(Trifluoromethoxy)phenylhydrazine hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: 397040050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

4-(Trifluoromethoxy)phenylhydrazine hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: 397040050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 397040050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
133115-72-7
IUPAC Name
[4-(trifluoromethoxy)phenyl]hydrazine hydrochloride
Molecular Formula
C7H8ClF3N2O
InChI Key
KQXZVSQCMVKMBK-UHFFFAOYSA-N
SMILES
Cl.NNC1=CC=C(OC(F)(F)F)C=C1
Appearance (Color)
White to light brown
Infrared spectrum
Conforms
Titration Argentometric
>=96.0 %
Appearance (Form)
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text