N-BOC-3-pyrrolidinone, 97%, Thermo Scientific Chemicals
N-BOC-3-pyrrolidinone, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-BOC-3-pyrrolidinone, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
1 g
Packaging:
Glass bottle
Catalog number 397050100
Price (USD)/ Each
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Nametert-butyl 3-oxopyrrolidine-1-carboxylate
Molecular FormulaC9H15NO3
InChI KeyJSOMVCDXPUXKIC-UHFFFAOYSA-N
SMILESCC(C)(C)OC(=O)N1CCC(=O)C1
Molecular Weight (g/mol)185.22
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light brown
Appearance (Form)Low melting solid
Infrared spectrumConforms
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only