Potassium Phosphate, Dibasic, 1.0M solution in water
Potassium Phosphate, Dibasic, 1.0M solution in water
Potassium Phosphate, Dibasic, 1.0M solution in water
Potassium Phosphate, Dibasic, 1.0M solution in water
Thermo Scientific Chemicals

Potassium Phosphate, Dibasic, 1.0M solution in water

CAS: 7758-11-4 | HK2O4P | 174.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 397080010
Price (USD)
46.65
Special offer
Online exclusive
Ends: 26-Sep-2025
57.60
Save 10.95 (19%)
Each
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
46.65
Special offer
Online exclusive
Ends: 26-Sep-2025
57.60
Save 10.95 (19%)
Each
Add to cart
Chemical Identifiers
CAS7758-11-4
IUPAC Namedipotassium hydrogen phosphate
Molecular FormulaHK2O4P
InChI KeyZPWVASYFFYYZEW-UHFFFAOYSA-L
SMILES[K+].[K+].OP([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Solution
Titration with HClO41.0 M Exact strength on the certificate of analysis
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Share catalog number, name or link