Thermo Scientific Chemicals

2,3-Pyrazinedicarboxylic anhydride, 97%, Thermo Scientific Chemicals

Catalog number: 397130250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

2,3-Pyrazinedicarboxylic anhydride, 97%, Thermo Scientific Chemicals

Catalog number: 397130250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 397130250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
4744-50-7
IUPAC Name
5H,7H-furo[3,4-b]pyrazine-5,7-dione
Molecular Formula
C6H2N2O3
InChI Key
AWJWCTOOIBYHON-UHFFFAOYSA-N
SMILES
O=C1OC(=O)C2=NC=CN=C12
Appearance (Color)
White to light brown
Appearance (Form)
Powder
Infrared spectrum
Conforms
Titration Acid
>=96 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text