Thermo Scientific Chemicals

3-(Bromomethyl)pyridine hydrobromide, 97%, Thermo Scientific Chemicals

Catalog number: 397140050
5 g, Each
Thermo Scientific Chemicals

3-(Bromomethyl)pyridine hydrobromide, 97%, Thermo Scientific Chemicals

Catalog number: 397140050
5 g, Each
Quantity
Catalog number: 397140050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
4916-55-6
IUPAC Name
hydrogen 3-(bromomethyl)pyridine bromide
Molecular Formula
C6H7Br2N
InChI Key
FNHPUOJKUXFUKN-UHFFFAOYSA-N
SMILES
[H+].[Br-].BrCC1=CC=CN=C1
Appearance (Form)
Powder or crystals and/or chunks
Infrared spectrum
Conforms
Melting point
148°C to 154°C
Titration with HClO4
>=96.0 %
Appearance (Color)
White to yellow to brown

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text