5-Formyl-2-thiopheneboronic acid, 97%, Thermo Scientific Chemicals
5-Formyl-2-thiopheneboronic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Formyl-2-thiopheneboronic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number 397150050
Price (USD)/ Each
274.00
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
274.00
Add to cart
5-Formyl-2-thiopheneboronic acid, 97%, Thermo Scientific Chemicals
Catalog number397150050
Price (USD)/ Each
274.00
-
Add to cart
Chemical Identifiers
CAS4347-33-5
IUPAC Name(5-formylthiophen-2-yl)boronic acid
Molecular FormulaC5H5BO3S
InChI KeyDEQOVKFWRPOPQP-UHFFFAOYSA-N
SMILESOB(O)C1=CC=C(S1)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to orange
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only