2-Methyltetrahydrofuran, 99+%, Extra Dry, stabilizer free, AcroSeal™, Thermo Scientific Chemicals
2-Methyltetrahydrofuran, 99+%, Extra Dry, stabilizer free, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Methyltetrahydrofuran, 99+%, Extra Dry, stabilizer free, AcroSeal™, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
Packaging:
AcroSeal™ Glass Bottle
Catalog number 397201000
Price (USD)/ Each
107.65
Online exclusive
119.00 
Save 11.35 (10%)
-
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (USD)/ Each
107.65
Online exclusive
119.00 
Save 11.35 (10%)
Add to cart
2-Methyltetrahydrofuran, 99+%, Extra Dry, stabilizer free, AcroSeal™, Thermo Scientific Chemicals
Catalog number397201000
Price (USD)/ Each
107.65
Online exclusive
119.00 
Save 11.35 (10%)
-
Add to cart
Chemical Identifiers
CAS96-47-9
IUPAC Name2-methyloxolane
Molecular FormulaC5H10O
InChI KeyJWUJQDFVADABEY-UHFFFAOYNA-N
SMILESCC1CCCO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
Water=<0.005 %
Residue after evaporation=<0.001 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only