2-Methylnicotinic acid, 98+%, Thermo Scientific Chemicals
2-Methylnicotinic acid, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Methylnicotinic acid, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Catalog number 397320250
Price (USD)/ Each
1,071.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
1,071.00
Add to cart
2-Methylnicotinic acid, 98+%, Thermo Scientific Chemicals
Catalog number397320250
Price (USD)/ Each
1,071.00
-
Add to cart
Chemical Identifiers
IUPAC Name2-methylpyridine-3-carboxylic acid
Molecular FormulaC7H7NO2
InChI KeyHNTZKNJGAFJMHQ-UHFFFAOYSA-N
SMILESCC1=C(C=CC=N1)C(O)=O
Molecular Weight (g/mol)137.14
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to light brown
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=98.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only