Thermo Scientific Chemicals

4-(Bromomethyl)pyridine hydrobromide, 97%, Thermo Scientific Chemicals

Catalog number: 397330050
5 g, Each
Thermo Scientific Chemicals

4-(Bromomethyl)pyridine hydrobromide, 97%, Thermo Scientific Chemicals

Catalog number: 397330050
5 g, Each
Quantity
Catalog number: 397330050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
73870-24-3
IUPAC Name
hydrogen 4-(bromomethyl)pyridine bromide
Molecular Formula
C6H7Br2N
InChI Key
VAJUUDUWDNCECT-UHFFFAOYSA-N
SMILES
[H+].[Br-].BrCC1=CC=NC=C1
Appearance (Color)
White to yellow
Appearance (Form)
Powder
Infrared spectrum
Conforms
Melting point
189°C to 192°C
Titration Argentometric
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text