Chemical Identifiers
CAS4747-71-1
IUPAC Name14,15-dimethyl-10-phenyl-2,11-diazatricyclo[10.4.0.0⁴,⁹]hexadeca-1(12),4,6,8,10,13,15-heptaen-3-one
Molecular FormulaC22H18N2O
InChI KeyUHJCHZGAXRAPFM-UHFFFAOYSA-N
SMILESCC1=CC2=C(C=C1C)N=C(C1=CC=CC=C1)C1=CC=CC=C1C(=O)N2
View more