Thermo Scientific Chemicals

4-(Trifluoromethyl)hydrocinnamic acid, 95%, Thermo Scientific Chemicals

Catalog number: 397380010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

4-(Trifluoromethyl)hydrocinnamic acid, 95%, Thermo Scientific Chemicals

Catalog number: 397380010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 397380010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
53473-36-2
IUPAC Name
3-[4-(trifluoromethyl)phenyl]propanoic acid
Molecular Formula
C10H9F3O2
InChI Key
OEIUMLSCWINLBB-UHFFFAOYSA-N
SMILES
OC(=O)CCC1=CC=C(C=C1)C(F)(F)F
Appearance (Form)
Crystalline powder
Appearance (Color)
White to light yellow
Infrared spectrum
Conforms
Melting point
111°C to 119°C
HPLC
>=94 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text