4-(Trifluoromethyl)hydrocinnamic acid, 95%, Thermo Scientific Chemicals
4-(Trifluoromethyl)hydrocinnamic acid, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-(Trifluoromethyl)hydrocinnamic acid, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Packaging:
Glass bottle
Catalog number 397380250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS53473-36-2
IUPAC Name3-[4-(trifluoromethyl)phenyl]propanoic acid
Molecular FormulaC10H9F3O2
InChI KeyOEIUMLSCWINLBB-UHFFFAOYSA-N
SMILESOC(=O)CCC1=CC=C(C=C1)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Appearance (Color)White to light yellow
Infrared spectrumConforms
Melting point111°C to 119°C
HPLC>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only