2,2-Dimethyl-1,3-dioxolane-4-methanamine, 97%, Thermo Scientific Chemicals
2,2-Dimethyl-1,3-dioxolane-4-methanamine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,2-Dimethyl-1,3-dioxolane-4-methanamine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 397410010
Price (USD)/ Each
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS22195-47-7
IUPAC Name1-(2,2-dimethyl-1,3-dioxolan-4-yl)methanamine
Molecular FormulaC6H13NO2
InChI KeyHXOYWCSTHVTLOW-UHFFFAOYNA-N
SMILESCC1(C)OCC(CN)O1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=96.0 %
Infrared spectrumConforms
Refractive index1.4370 to 1.4390 (20°C, 589 nm)
Appearance (Color)Colorless to yellow
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only