Cyclopentane, 95+%, pure, Thermo Scientific Chemicals
Cyclopentane, 95+%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cyclopentane, 95+%, pure, Thermo Scientific Chemicals

Cyclopentane, 95+%, C5H10, CAS Number-287-92-3, pentamethylene, cyclopentanes, hsdb 62, hydrocarbons, cyclic c5 and c6, cyclopentan, t86pb90rnu, unii-t86pb90rnu, ciclopentano, cyclopentyl group, zyklopentan, 5L, 95% min. (GC), 50.0 deg.C, CHEBI:23492, Colorless, 0.7500g/mL, 206-016-6, -7 deg.C
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 L
1 L
Catalog number 397430050
Price (USD)/ Each
-
Quantity:
5 L
Request bulk or custom format
Chemical Identifiers
CAS287-92-3
IUPAC Namecyclopentane
Molecular FormulaC5H10
InChI KeyRGSFGYAAUTVSQA-UHFFFAOYSA-N
SMILESC1CCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=95.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only