1-Acetylguanidine, 97%, Thermo Scientific Chemicals
1-Acetylguanidine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Acetylguanidine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 397440050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC NameN-(diaminomethylidene)acetamide
Molecular FormulaC3H7N3O
InChI KeyNGGXACLSAZXJGM-UHFFFAOYSA-N
SMILESCC(=O)N=C(N)N
Molecular Weight (g/mol)101.11
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only