1-Acetylguanidine, 97%
1-Acetylguanidine, 97%
1-Acetylguanidine, 97%
1-Acetylguanidine, 97%
Thermo Scientific Chemicals

1-Acetylguanidine, 97%

CAS: 5699-40-1 | C3H7N3O | 101.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 397440250
Price (EUR)
191,25
Online Exclusive
225,00
Save 33,75 (15%)
Each
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
191,25
Online Exclusive
225,00
Save 33,75 (15%)
Each
Add to cart
Chemical Identifiers
CAS5699-40-1
IUPAC NameN-(diaminomethylidene)acetamide
Molecular FormulaC3H7N3O
InChI KeyNGGXACLSAZXJGM-UHFFFAOYSA-N
SMILESCC(=O)N=C(N)N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Share catalog number, name or link