Magnesium ethoxide, 98%, Thermo Scientific Chemicals
Magnesium ethoxide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Magnesium ethoxide, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
500 g
Packaging:
Glass bottle
Catalog number 397471000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2414-98-4
IUPAC Namemagnesium(2+) diethanolate
Molecular FormulaC4H10MgO2
InChI KeyXDKQUSKHRIUJEO-UHFFFAOYSA-N
SMILES[Mg++].CC[O-].CC[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HCl97.0 to 103.0 %
Appearance (Color)White to light grey
Infrared spectrumConforms
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only