Thermo Scientific Chemicals

Sarcosine methyl ester, hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: 397502500
250 g, Glass bottle, Each
Thermo Scientific Chemicals

Sarcosine methyl ester, hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: 397502500
250 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 397502500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13515-93-0
Appearance (Color)
White to beige
Appearance (Form)
Crystalline powder or crystals
Infrared spectrum
Conforms
Titration with HClO4
>=96.0 %
Loss on drying
=<1.0 % (1 g, 70°C) (to constant weight)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text