Iodopyrazine, 97%, Thermo Scientific Chemicals
Iodopyrazine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Iodopyrazine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 397530050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS32111-21-0
IUPAC Name2-iodopyrazine
Molecular FormulaC4H3IN2
InChI KeyOYWPFIUVDKHHGQ-UHFFFAOYSA-N
SMILESIC1=CN=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to brown
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=96.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only