2,6-Dimethylbenzaldehyde, 97%, Thermo Scientific Chemicals
2,6-Dimethylbenzaldehyde, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,6-Dimethylbenzaldehyde, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 397560010
Price (USD)/ Each
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1123-56-4
IUPAC Name2,6-dimethylbenzaldehyde
Molecular FormulaC9H10O
InChI KeyQOJQBWSZHCKOLL-UHFFFAOYSA-N
SMILESCC1=CC=CC(C)=C1C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to yellow
Appearance (Form)Liquid or low melting solid
Infrared spectrumConforms
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only