Thermo Scientific Chemicals

(Trimethylsilyl)diazomethane, approx. 2M solution in diethyl ether, Thermo Scientific Chemicals

Catalog number: 397620250
25 mL, Glass bottle, Each
Thermo Scientific Chemicals

(Trimethylsilyl)diazomethane, approx. 2M solution in diethyl ether, Thermo Scientific Chemicals

Catalog number: 397620250
25 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 397620250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
18107-18-1
IUPAC Name
(diazomethyl)trimethylsilane
Molecular Formula
C4H10N2Si
InChI Key
ONDSBJMLAHVLMI-UHFFFAOYSA-N
SMILES
C[Si](C)(C)C=[N+]=[N-]
Appearance (Color)
Yellow
Appearance (Form)
Solution
Appearance
May develop some turbidity or precipitate
Appearance
If precipitation is present, gently mix
Appearance
precipitate back into solution under an inert

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text