RUO – Research Use Only
Please enter sku to load default questions
Frequently asked questions (FAQs)
What type of glass are the Glass Beads, 500 to 750um made out of?
The Glass Beads, 500 to 750µm are made of soda lime glass.