4-Fluoroindole, 96%, Thermo Scientific Chemicals
4-Fluoroindole, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Fluoroindole, 96%, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
1 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 397650010
Price (USD)/ Each
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS387-43-9
IUPAC Name4-fluoro-1H-indole
Molecular FormulaC8H6FN
InChI KeyZWKIJOPJWWZLDI-UHFFFAOYSA-N
SMILESFC1=C2C=CNC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to beige
Appearance (Form)Low melting solid
Infrared spectrumConforms
GC>=95 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only