Thermo Scientific Chemicals

tert-Butyl diethylphosphonoacetate, 95%, Thermo Scientific Chemicals

Catalog number: 397750050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

tert-Butyl diethylphosphonoacetate, 95%, Thermo Scientific Chemicals

Catalog number: 397750050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 397750050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
27784-76-5
IUPAC Name
tert-butyl 2-(diethoxyphosphoryl)acetate
Molecular Formula
C10H21O5P
InChI Key
NFEGNISFSSLEGU-UHFFFAOYSA-N
SMILES
CCOP(=O)(CC(=O)OC(C)(C)C)OCC
Appearance (Form)
Liquid
Appearance (Color)
Clear colorless
Infrared spectrum
Conforms
GC
>=94 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for C-C Bond Formation (Olefination)
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text