Thermo Scientific Chemicals

3,3,3-Trifluoro-2-(trifluoromethyl)propionic acid, 97%, Thermo Scientific Chemicals

Catalog number: 397860050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

3,3,3-Trifluoro-2-(trifluoromethyl)propionic acid, 97%, Thermo Scientific Chemicals

Catalog number: 397860050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 397860050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
564-10-3
IUPAC Name
3,3,3-trifluoro-2-(trifluoromethyl)propanoic acid
Molecular Formula
C4H2F6O2
InChI Key
RAEAYTICAPHWJW-UHFFFAOYSA-N
SMILES
OC(=O)C(C(F)(F)F)C(F)(F)F
Appearance (Color)
White
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Melting point
49°C to 53°C
Titration with NaOH
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text