Milrinone, 97%
Thermo Scientific Chemicals

Milrinone, 97%

Have Questions?
Quantity:
50 mg
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 397870500
Price (USD)/ Each
-
Quantity:
50 mg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS78415-72-2
IUPAC Name2-methyl-6-oxo-1,6-dihydro-[3,4'-bipyridine]-5-carbonitrile
Molecular FormulaC12H9N3O
InChI KeyPZRHRDRVRGEVNW-UHFFFAOYSA-N
SMILESCC1=C(C=C(C#N)C(=O)N1)C1=CC=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light pink
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only