Thermo Scientific Chemicals

Methyl indole-6-carboxylate, 97%, Thermo Scientific Chemicals

Catalog number: 397930050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Methyl indole-6-carboxylate, 97%, Thermo Scientific Chemicals

Catalog number: 397930050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 397930050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
50820-65-0
IUPAC Name
methyl 1H-indole-6-carboxylate
Molecular Formula
C10H9NO2
InChI Key
AYYOZKHMSABVRP-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC=C2C=CNC2=C1
GC
>=96.0 %
Appearance (Color)
White to light brown
Appearance (Form)
Powder
Infrared spectrum
Conforms
Melting point
79°C to 84°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text