2-Nitroethanol, 97%
2-Nitroethanol, 97%
2-Nitroethanol, 97%
Thermo Scientific Chemicals

2-Nitroethanol, 97%

CAS: 625-48-9 | C2H5NO3 | 91.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 397960050
Price (EUR)
40,12
Online Exclusive
47,20
Save 7,08 (15%)
Each
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
40,12
Online Exclusive
47,20
Save 7,08 (15%)
Each
Add to cart
Chemical Identifiers
IUPAC Name2-nitroethan-1-ol
Molecular FormulaC2H5NO3
InChI KeyKIPMDPDAFINLIV-UHFFFAOYSA-N
SMILESOCC[N+]([O-])=O
Molecular Weight (g/mol)91.07
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=96.0 %
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4430 to 1.4450 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow to orange
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Share catalog number, name or link