Thermo Scientific Chemicals

(1S,4S)-(-)-2-BOC-2,5-diazabicyclo[2.2.1]heptane, 95%, Thermo Scientific Chemicals

Catalog number: 398040010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

(1S,4S)-(-)-2-BOC-2,5-diazabicyclo[2.2.1]heptane, 95%, Thermo Scientific Chemicals

Catalog number: 398040010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 398040010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
113451-59-5
IUPAC Name
tert-butyl 2,5-diazabicyclo[2.2.1]heptane-2-carboxylate
Molecular Formula
C10H18N2O2
InChI Key
UXAWXZDXVOYLII-UHFFFAOYNA-N
SMILES
CC(C)(C)OC(=O)N1CC2CC1CN2
Melting point
73°C to 77°C
GC
>=94 %
Appearance (Color)
White to light beige
Infrared spectrum
Conforms
Specific optical rotation
-41° to -47° (20°C, 589 nm) (C=1, chloroform)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text