Thermo Scientific Chemicals

3-Aminopiperidine dihydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: 398110010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

3-Aminopiperidine dihydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: 398110010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 398110010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
138060-07-8
IUPAC Name
piperidin-3-amine dihydrochloride
Molecular Formula
C5H14Cl2N2
InChI Key
GGPNYXIOFZLNKW-UHFFFAOYNA-N
SMILES
Cl.Cl.NC1CCCNC1
Appearance (Color)
Off-white to beige
Appearance (Form)
Powder or solid
Infrared spectrum
Conforms
Titration with HClO4
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text