Thermo Scientific Chemicals

BOC-L-α-phenylglycine, 99%, Thermo Scientific Chemicals

Catalog number: 398160050
5 g, Glass Bottle, Each
Thermo Scientific Chemicals

BOC-L-α-phenylglycine, 99%, Thermo Scientific Chemicals

Catalog number: 398160050
5 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 398160050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2900-27-8
IUPAC Name
2-{[(tert-butoxy)carbonyl]amino}-2-phenylacetic acid
Molecular Formula
C13H17NO4
InChI Key
HOBFSNNENNQQIU-UHFFFAOYNA-N
SMILES
CC(C)(C)OC(=O)NC(C(O)=O)C1=CC=CC=C1
Melting point
88°C to 91°C
Titration with NaOH
>=98.5 %
Specific optical rotation
+142° to +146° (20°C, 589 nm) (c=1%, ethanol)
Appearance (Color)
White to off-white
Appearance (Form)
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text