Thermo Scientific Chemicals

2-Hydroxy-5-(trifluoromethoxy)benzaldehyde, 99%, Thermo Scientific Chemicals

Catalog number: 398200010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

2-Hydroxy-5-(trifluoromethoxy)benzaldehyde, 99%, Thermo Scientific Chemicals

Catalog number: 398200010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 398200010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
93249-62-8
IUPAC Name
2-hydroxy-5-(trifluoromethoxy)benzaldehyde
Molecular Formula
C8H5F3O3
InChI Key
WQUZBERVMUEJTD-UHFFFAOYSA-N
SMILES
OC1=C(C=O)C=C(OC(F)(F)F)C=C1
Infrared spectrum
Conforms
NMR (1H-NMR)
Consistent with structure
Melting point
30°C to 34°C
HPLC
>=98.5 %
Appearance (Color)
White to light yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text