Thermo Scientific Chemicals

2,4-Dichloro-5-methylpyrimidine, 98%, Thermo Scientific Chemicals

Catalog number: 398290010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

2,4-Dichloro-5-methylpyrimidine, 98%, Thermo Scientific Chemicals

Catalog number: 398290010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 398290010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1780-31-0
IUPAC Name
2,4-dichloro-5-methylpyrimidine
Molecular Formula
C5H4Cl2N2
InChI Key
DQXNTSXKIUZJJS-UHFFFAOYSA-N
SMILES
CC1=CN=C(Cl)N=C1Cl
Infrared spectrum
Conforms
GC
>=97.5 %
Appearance (Color)
Light yellow
Appearance (Form)
Low melting solid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text