Isocytosine, 99%, Thermo Scientific Chemicals
Isocytosine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Isocytosine, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 398320050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS108-53-2
IUPAC Name2-amino-3,4-dihydropyrimidin-4-one
Molecular FormulaC4H5N3O
InChI KeyXQCZBXHVTFVIFE-UHFFFAOYSA-N
SMILESNC1=NC=CC(=O)N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=98.5 %
Solubility(50 mg/ml in DMSO) Clear solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only