2-chloro-5-cyanopyridine, 97%, Thermo Scientific Chemicals
2-chloro-5-cyanopyridine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-chloro-5-cyanopyridine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 398390050
Price (USD)/ Each
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS33252-28-7
IUPAC Name6-chloropyridine-3-carbonitrile
Molecular FormulaC6H3ClN2
InChI KeyORIQLMBUPMABDV-UHFFFAOYSA-N
SMILESClC1=CC=C(C=N1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to beige
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point116°C to 120°C
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only