2-Amino-m-cresol, 96%, Thermo Scientific Chemicals
2-Amino-m-cresol, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Amino-m-cresol, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 398410050
Price (USD)/ Each
82.40
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
82.40
Add to cart
2-Amino-m-cresol, 96%, Thermo Scientific Chemicals
Catalog number398410050
Price (USD)/ Each
82.40
-
Add to cart
Chemical Identifiers
CAS2835-97-4
IUPAC Name2-amino-3-methylphenol
Molecular FormulaC7H9NO
InChI KeyFEDLEBCVFZMHBP-UHFFFAOYSA-N
SMILESCC1=CC=CC(O)=C1N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to brown
Appearance (Form)Crystalline powder or flakes
Infrared spectrumConforms
Melting point149°C to 152°C
HPLC>=95 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only