4-(Trifluoromethyl)styrene, 98+%, stabilized with 0.1% 4-tert-butylcatechol, Thermo Scientific Chemicals
4-(Trifluoromethyl)styrene, 98+%, stabilized with 0.1% 4-tert-butylcatechol, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-(Trifluoromethyl)styrene, 98+%, stabilized with 0.1% 4-tert-butylcatechol, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
1 g
Packaging:
Glass bottle
Catalog number 398460010
Price (USD)/ Each
136.00
-
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
136.00
Add to cart
4-(Trifluoromethyl)styrene, 98+%, stabilized with 0.1% 4-tert-butylcatechol, Thermo Scientific Chemicals
Catalog number398460010
Price (USD)/ Each
136.00
-
Add to cart
Chemical Identifiers
IUPAC Name1-ethenyl-4-(trifluoromethyl)benzene
Molecular FormulaC9H7F3
InChI KeyCEWDRCQPGANDRS-UHFFFAOYSA-N
SMILESFC(F)(F)C1=CC=C(C=C)C=C1
Molecular Weight (g/mol)172.15
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4650 to 1.4680 (20°C, 589 nm)
GC>=98.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only