Thermo Scientific Chemicals

Imidazole-2-carboxaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: 398470050
5 g, Each
Thermo Scientific Chemicals

Imidazole-2-carboxaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: 398470050
5 g, Each
Quantity
Catalog number: 398470050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
10111-08-7
IUPAC Name
1H-imidazole-2-carbaldehyde
Molecular Formula
C4H4N2O
InChI Key
XYHKNCXZYYTLRG-UHFFFAOYSA-N
SMILES
O=CC1=NC=CN1
HPLC
>=97.5 %
Appearance (Color)
Off-white to light tan
Appearance (Form)
Powder
Infrared spectrum
Conforms
NMR (1H-NMR)
Consistent with structure

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text