Thermo Scientific Chemicals

4-Maleimidobutyric acid N-hydroxysuccinimide ester, 97%, Thermo Scientific Chemicals

Catalog number: 398681000
100 mg, Glass bottle, Each
Thermo Scientific Chemicals

4-Maleimidobutyric acid N-hydroxysuccinimide ester, 97%, Thermo Scientific Chemicals

Catalog number: 398681000
100 mg, Glass bottle, Each
Quantity
Packaging
Catalog number: 398681000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
80307-12-6
IUPAC Name
2,5-dioxopyrrolidin-1-yl 4-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)butanoate
Molecular Formula
C12H12N2O6
InChI Key
PVGATNRYUYNBHO-UHFFFAOYSA-N
SMILES
O=C(CCCN1C(=O)C=CC1=O)ON1C(=O)CCC1=O
Appearance (Color)
White to light yellow
Appearance (Form)
Powder
Infrared spectrum
Conforms
Melting point
123°C to 130°C
HPLC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text